GC Analysis of 1-Cyclohexylethylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® B-DP
GC Analysis of 1-Cyclohexylethylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® B-DP
Conditions
column Astec CHIRALDEX B-DP, 30 m x 0.25 mm I.D., 0.12 µm (78023AST)
oven 130 °C
carrier gas helium, 30 psi
sample peak 1: S(+)-cyclohexylethylamine (N-TFA derivative)peak 2: R(-)-cyclohexylethylamine (N-TFA derivative)
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Amines, Enantiomers
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes