GC Analysis of 1-Methoxy-2-Propylamine Enantiomers (N-Chloroacetyl Derivatives) on Astec® CHIRALDEX® B-PH
GC Analysis of 1-Methoxy-2-Propylamine Enantiomers (N-Chloroacetyl Derivatives) on Astec® CHIRALDEX® B-PH
Conditions
column Astec CHIRALDEX B-PH, 30 m x 0.25 mm I.D., 0.12 µm (71023AST)
oven 120 °C
carrier gas helium, 24 psi
sample 3 mg/mL in methanolpeaks 1 & 2: 1-methoxy-2-propylamine enantiomers (N-chloroacetyl derivatives)
injection 1 µL, 80:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Amines
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes