GC Analysis of 1-Phenylethanol Enantiomers on Astec® CHIRALDEX® B-PM
GC Analysis of 1-Phenylethanol Enantiomers on Astec® CHIRALDEX® B-PM
Conditions
column Astec CHIRALDEX B-PM, 30 m x 0.25 mm I.D., 0.12 µm (76023AST)
oven 120 C
carrier gas helium, 24 psi
sample 3 mg/mL in methanolpeak 1: R(+)-1-phenylethanolpeak 2: S(-)-1-phenylethanol
injection 1 µL, 80:1 split
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Alcohols
Featured Industry Chemicals and Industrial Polymers
Flavors and Fragrances
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes