GC Analysis of 1-Phenylethylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® G-PN
GC Analysis of 1-Phenylethylamine Enantiomers (N-Trifluoroacetyl Derivatives) on Astec® CHIRALDEX® G-PN
Conditions
column Astec CHIRALDEX G-PN, 30 m x 0.25 mm I.D., 0.12 µm (74033AST)
oven 130 °C
carrier gas helium, 30 psi
sample 3 mg/mL in methanolpeak 1: R(+)-1-phenylethylamine (N-TFA derivative)peak 2: S(-)-1-phenylethylamine (N-TFA derivative)
injection 1 µL, 80:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Can reverse elution order by using N-acetyl derivative on CHIRALDEX B-DM and B-PM, the N-chloroacetyl derivative on B-PM, or the N-TFA derivative on B-PH.
Categories Analytical Chromatography, GC Applications, Anilines, Amines, Enantiomers
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes