GC Analysis of 2,5-Dimethoxytetrahydrofuran Enantiomers on Astec® CHIRALDEX® G-BP
GC Analysis of 2,5-Dimethoxytetrahydrofuran Enantiomers on Astec® CHIRALDEX® G-BP
Conditions
column Astec CHIRALDEX G-BP, 20 m x 0.25 mm I.D., 0.12 µm (75032AST)
oven 60 °C
carrier gas helium, 20 psi
injection 0.2 µL, 80:1 split
sample gas 2 mg/mL in ethanolpeaks 1 & 2: 2,5-dimethoxytetrahydrofuran enantiomerspeak 3: meso-isomer
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Ethers, Enantiomers
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes