GC Analysis of 2,5-Dimethoxytetrahydrofuran Enantiomers on Astec® CHIRALDEX® G-TA
GC Analysis of 2,5-Dimethoxytetrahydrofuran Enantiomers on Astec® CHIRALDEX® G-TA
Conditions
column Astec CHIRALDEX G-TA, 30 m x 0.25 mm I.D., 0.12 µm (73033AST)
oven 70 °C
carrier gas helium, 30 psi
sample 2 mg/mL in ethanolpeaks 1 & 2: 2,5-dimethoxytetrahydrofuran enantiomerspeak 3: meso-isomer
injection 0.2 µL, 80:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Enantiomers, Ethers
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes