GC Analysis of 2-Hydroxycaproic Acid Methyl Ester Enantiomers on Astec® CHIRALDEX® B-DP
GC Analysis of 2-Hydroxycaproic Acid Methyl Ester Enantiomers on Astec® CHIRALDEX® B-DP
Conditions
column Astec CHIRALDEX B-DP, 30 m x 0.25 mm I.D., 0.12 µm (78023AST)
oven 100 °C
carrier gas helium, 30 psi
sample peaks 1 & 2: 2-hydroxycaproic acid methyl ester enantiomers
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Chiral Applications, Clinical, Forensic & Toxicology, Acids, Enantiomers, Biomarkers
Featured Industry Chemicals and Industrial Polymers
Clinical
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes