GC Analysis of 4-Methyloctanoic Acid Enantiomers on γ-DEX™ 120
GC Analysis of 4-Methyloctanoic Acid Enantiomers on γ-DEX™ 120
Conditions
column gamma-DEX 120, 30 m x 0.25 mm I.D., 0.25 µm (24307)
oven 115 °C
carrier gas helium, 20 cm/sec
sample ~1 mg/mL each enantiomer in methylene chloride
injection 1 µL, 100:1 split
inj. temp. 220 °C
detector FID, 300 °C
Description
Categories Analytical Chromatography, GC Applications, Organic Acids, Enantiomers, Fatty Acids
Featured Industry Chemicals and Industrial Polymers
Food and Beverages
Legal Information DEX is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes