GC Analysis of 5-Amino-1,3,3-Trimethylcyclohexanemethylamine Enantiomers (N-TFA Derivatives) on Astec® CHIRALDEX® B-TA
GC Analysis of 5-Amino-1,3,3-Trimethylcyclohexanemethylamine Enantiomers (N-TFA Derivatives) on Astec® CHIRALDEX® B-TA
Conditions
column Astec CHIRALDEX B-TA, 30 m x 0.25 mm I.D., 0.12 µm (73023AST)
oven 160 C
carrier gas helium, 30 psi
sample 3 mg/mL racemate in methanolpeaks 1 & 2: first enantiomer pairpeaks 3 & 4: second enantiomer pair
injection 1 µL, 80:1 split
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Amines, Enantiomers
Featured Industry Chemicals and Industrial Polymers
Legal Information Astec is a registered trademark of Sigma-Aldrich Co. LLC, CHIRALDEX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes