Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of a 10-Component Glyceride Standard (T6-T60 Saturates) on SLB®-35ms
GC Analysis of a 10-Component Glyceride Standard (T6-T60 Saturates) on SLB®-35ms
Conditions
column SLB-35ms, 10 m x 0.25 mm I.D., 0.10 µm; made by cutting down a 30 m x 0.25 mm I.D., 0.10 µm column (29802-U)
oven 75 C (0.1 min), 15 C/min to 380 C (10 min)
carrier gas helium, 1.5 ml/min
sample 10-component triglyceride standard (T6-T60 saturates)
injection 1muL, on-column
detector FID, 380 C
Description
Analysis Note Triglycerides are the main constituent of vegetable oil and animal fat, and make up most of the fats digested by humans. Most natural fats contain a complex mixture of individual triglycerides. Therfore, efficient capillary GC columns are required for ample resolution and subsequent identification. Triglycerides are large compounds, and require a relatively high final oven temperature for GC analysis. A short, thin film version of SLB-35ms (cut-down from a longer column) run to a final oven temperature of 380 ºC was found to provide good resolution and reasonable elution times.
Categories Analytical Chromatography, GC Applications, Glycerides
Featured Industry Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC