GC Analysis of a 12-Component Chlorinated Solvents Mix on SLB®-IL60, Comparison to a Polyethylene Glycol (PEG) Column
GC Analysis of a 12-Component Chlorinated Solvents Mix on SLB®-IL60, Comparison to a Polyethylene Glycol (PEG) Column
Conditions
column SLB-IL60, 30 m x 0.25 mm I.D., 0.20 µm (29505-U)
column PEG 2, 30 m x 0.25 mm I.D., 0.25 µm
oven 40 °C (4 min), 8 °C/min to 200 °C (5 min)
carrier gas helium, 30 cm/sec
sample 12-component chlorinated solvent mix, each analyte at 0.2% (v/v) in pentane
injection 1 µL100:1 split
liner 4 mm I.D., split/splitless type, single taper wool packed FocusLiner™ design
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons, Solvents
Featured Industry Chemicals and Industrial Polymers
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 1,1,1,2-Tetrachloroethane
  • 1,1,1-Trichloroethane
  • 1,1,2,2-Tetrachloroethane
  • 1,1-Dichloroethane
  • 1,1-Dichloroethylene
  • 1,2-Dichloroethane
  • Carbon tetrachloride
  • Chloroform
  • Methylene chloride
  • Tetrachloroethene
  • trans-1,2-Dichloroethylene
  • Trichloroethylene