Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of a 12-Component Esters and Ethers Mix on SLB®-IL60, Comparison to a Polyethylene Glycol (PEG) Column
GC Analysis of a 12-Component Esters and Ethers Mix on SLB®-IL60, Comparison to a Polyethylene Glycol (PEG) Column
Conditions
column PEG, 30 m x 0.25 mm I.D., 0.25 µm
column SLB-IL60, 30 m x 0.25 mm I.D., 0.20 µm (29505-U)
oven 40 C (4 min), 8 C/min to 200 C (5 min)
carrier gas helium, 30 cm/sec
sample 12-component esters and ethers mix, each analyte at 0.2% (v/v) in pentane
injection 1 muL100:1 split
liner 4 mm I.D., split/splitless type, single taper wool packed FocusLiner(TM) design
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Ethers, Esters
Featured Industry Chemicals and Industrial Polymers
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 1,4-Dioxane
  • Ethyl acetate
  • Ethyl formate
  • Isoamyl acetate
  • Isobutyl acetate
  • Isopropyl acetate
  • Methyl acetate
  • Methyl formate
  • n-Amyl acetate
  • n-Butyl acetate
  • n-Propyl acetate
  • Tetrahydrofuran (THF)