GC Analysis of a 14-Component Polybrominated Diphenyl Ether (PBDE) Mix on SLB®-5ms
GC Analysis of a 14-Component Polybrominated Diphenyl Ether (PBDE) Mix on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 125 °C (1 min), 10 °C/min to 340 °C (15 min)
carrier gas helium, 1.5 mL/min, constant
sample PBDE standard, 2.5-10 µg/mL in n-nonane
injection 1 µL, splitless, pulsed (30 psi until 0.2 min)
liner 4 mm I.D. single taper
inj. temp. 300 °C
MSD interface 340 °C
scan range SIM
Description
Categories Analytical Chromatography, GC Applications, Ethers, Dioxins/PCBs/PBDEs
Featured Industry Chemicals and Industrial Polymers
Environmental
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes