GC Analysis of a 16-Component Anilines Mix on SLB®-IL59
GC Analysis of a 16-Component Anilines Mix on SLB®-IL59
Conditions
column SLB-IL59, 30 m x 0.25 mm I.D., 0.20 mm (28891-U)
oven 75 °C, (2 min) 10 °C/min to 200 °C, 20 °C/min to 300 °C (10 min)
carrier gas helium, 1.5 mL/min constant
sample Each analyte at 5000 µg/mL in methylene chloride
injection 0.5 µL100:1 split
liner 4 mm I.D., split/splitless type, single taper design
inj. temp. 250 °C
detector FIID, 300 °C
Description
Categories Analytical Chromatography, GC Applications, Anilines
Featured Industry Chemicals and Industrial Polymers
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 2,4,5-Trichloroaniline
  • 2,4-Diaminotoluene
  • 2,6-Diethylaniline
  • 2,6-Dimethylaniline
  • 2-Chloroaniline
  • 2-Methyl-6-ethylaniline
  • 3,4-Dichloroaniline
  • 3-Aminobenzotrifluoride
  • 3-Chloro-4-fluoroaniline
  • 3-Chloro-4-methylaniline
  • 4-Chloro-2-nitroaniline
  • 4-Isopropylaniline
  • Aniline
  • Dimethylaniline
  • o-Toluidine