GC Analysis of a 17-Component Dioxin Congener Mix on SLB®-5ms
GC Analysis of a 17-Component Dioxin Congener Mix on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 150 °C (1 min), 5 °C/min to 325 °C (2 min)
carrier gas helium, 37 cm/sec, constant
sample dioxane/furan standard, 100-500 ppb in n-nonane
injection 1 µL, splitless (1 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
MSD interface 325 °C
scan range SIM
Description
Categories Analytical Chromatography, GC Applications, Dioxins/PCBs/PBDEs
Featured Industry Agriculture
Environmental
Food and Beverages
Industrial Hygiene
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes