GC Analysis of a 17-Component Polynuclear Aromatic Hydrocarbon (PAH) Mix on SLB®-IL59
GC Analysis of a 17-Component Polynuclear Aromatic Hydrocarbon (PAH) Mix on SLB®-IL59
Conditions
column SLB-IL59, 20 m x 0.18 mm I.D., 0.14 µm (custom)
oven 65 ºC (0.5 min), 25 ºC/min to 300 ºC (20 min)
carrier gas helium, 50 psi constant pressure
sample each PAH at 10 µg/mL in methylene chloride
injection 1 µLsplitless (1 min)
liner 4 mm I.D., split/splitless type, wool packed single taper FocusLiner™ design
inj. temp. 265 °C
detector MSD, interface at 300 ºC, m/z = 50-500
Description
Categories Analytical Chromatography, GC Applications, PAHs, Hydrocarbons
Featured Industry Environmental
Food and Beverages
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • Acenaphthene
  • Acenaphthylene
  • Anthracene
  • Benzo[a]anthracene
  • Benzo[a]pyrene
  • Benzo[b]fluoranthene
  • Benzo[g,h,i]perylene
  • Benzo[j]fluoranthene
  • Benzo[k]fluoranthene
  • Chrysene
  • Dibenzo[a,h]anthracene
  • Fluoranthene
  • Fluorene
  • Indeno[1,2,3-cd]pyrene
  • Naphthalene
  • Phenanthrene
  • Pyrene