GC Analysis of a 19-Component PCB Congener Mix on SP™-2331
GC Analysis of a 19-Component PCB Congener Mix on SP™-2331
Conditions
column SP-2331, 30 m x 0.25 mm I.D., 0.20 µm (24257)
oven 60 °C (1 min), 8 °C/min to 230 °C
carrier gas helium, 1.5 mL/min constant
sample PCB congener mix, each analyte at 2.5 ppm in n-hexane
injection 1 µLsplitless (1.0 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
MSD interface 220 °C
scan range m/z = 35-500
Description
Categories Analytical Chromatography, GC Applications, Dioxins/PCBs/PBDEs
Featured Industry Environmental
Food and Beverages
Legal Information SP is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes