Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of a 20-Component Organochlorine Pesticide Mix in Water on a 5% Phenyl Column after SPE using Supelclean™ ENVI™-8
Conditions
sample preparation SPE (Solid Phase Extraction)
sample/matrix waste water spiked with pesticides
SPE tube/cartridge Supelclean ENVI-8, glass hardware, 500 mg/6 mL (57107 (500 mg/6 mL); 57108 (1 g/6 mL))
condition 3 mL methanol; h 2 mL 5% methanol in water
sample addition load sample at 5 ml/min
drying 2-3 min with nitrogen stream
elution 2 x 4 mL hexane:acetone (90:10), drop wise. allow to soak into bed
column 5% Phenyl Polysiloxane, 15 m x 0.53 mm I.D., 0.50 µm (Supelco equivalent, Equity-5, 28252-U)
oven 150 C (0.5 min) to 275 C (5 min)
carrier gas helium
injection 1 muLsplit/splitless (45 sec delay)
inj. temp. 200 C
detector ECD, 300 C
Description
Application Sample Pre-treatment:
Add 5 mL methanol to 100 mL aqueous hazardous waste. Adjust to pH 5-7 if necessary.
Categories Analytical Chromatography, SPE Applications, GC Applications, Pesticides
Featured Industry Environmental
Food and Beverages
Legal Information ENVI is a trademark of Sigma-Aldrich Co. LLC, Supelclean is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC, application for SPE
Analyte
  • 1,1,1-Trichloro-2,2-bis(4-chlorophenyl)ethane (4,4′-DDT)
  • 1,1-Dichloro-2,2-bis(4-chlorophenyl)ethane (4,4′-DDD)
  • 4,4′-DDE (1,1-Dichloro-2,2-bis(p-chlorophenyl)ethylene)
  • Aldrin
  • Decachlorobiphenyl
  • Dieldrin
  • Endosulfan I
  • Endosulfan II
  • Endosulfan sulfate
  • Endrin
  • Endrin aldehyde
  • Endrin ketone
  • Heptachlor
  • Heptachlor epoxide
  • Methoxychlor
  • α-BHC
  • α-Chlordane
  • β-BHC
  • γ-BHC
  • γ-Chlordane
  • δ-BHC