GC Analysis of a 32-Component Anilines Mix on SLB®-IL59
GC Analysis of a 32-Component Anilines Mix on SLB®-IL59
Conditions
column SLB-IL59, 30 m x 0.25 mm I.D., 0.20 µm (28891-U)
oven 75 °C (2 min), 10 °C/min to 250 °C, 15 °C/min to 300 °C (25 min)
carrier gas helium, 0.7 mL/min constant
sample Each analyte at 50 µg/mL in methylene chloride
injection 1 µLpulsed splitless (25 psi pulse)
liner 4 mm I.D., split/splitless type, single taper design
inj. temp. 250 °C
detector MSD, interface 275 °C, m/z = 30-300
Description
Categories Analytical Chromatography, GC Applications, Anilines
Featured Industry Chemicals and Industrial Polymers
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 2,4,5-Trimethylaniline
  • 2,4-Dimethylaniline
  • 2,6-Dimethylaniline
  • 2-Aminoanthraquinone
  • 2-Aminotoluene
  • 2-Methoxy-5-methylaniline
  • 2-Methoxy-5-nitroaniline
  • 2-Nitroaniline
  • 3-(Chloromethyl)pyridine
  • 3,3’-Dichlorobenzidine
  • 3,3’-Dimethoxybenzidine
  • 3,3’-Dimethylbenzidine
  • 3-Aminotoluene
  • 3-Nitroaniline
  • 4,4’-Methylene-bis-(o-chloroaniline)
  • 4,4’-Methylenedianiline
  • 4,4’-Oxydianiline
  • 4,4'-Methylene-bis(N,N-dimethylaniline)
  • 4-Aminobiphenyl
  • 4-Aminotoluene
  • 4-Chloro-2-methylaniline
  • 4-Chloroaniline
  • 4-Nitroaniline
  • 5-Nitro-o-toluidine
  • Aminoazobenzene
  • Aniline
  • Azobenzene
  • Benzidine
  • Dimethylaminoazobenzene
  • Diphenylamine
  • α-Anisidine
  • β-Naphthylamine