GC Analysis of a 37-Component FAME Mix on SP™-2560, Slower Oven Ramp
GC Analysis of a 37-Component FAME Mix on SP™-2560, Slower Oven Ramp
Conditions
column SP-2560, 100 m x 0.25 mm I.D., 0.20 µm (24056)
oven 140 C (5 min) to 240 C (5 min) at 4 C/min
carrier gas helium, 20 cm/sec
sample volume 1 muL split 100:1, 260 C
detector FID, 260 C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information SP is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes