GC Analysis of a 37-Component FAME Mix on SPB-PUFA
GC Analysis of a 37-Component FAME Mix on SPB-PUFA
Conditions
column SPB-PUFA, 30 m x 0.32 mm I.D., 0.20 µm (24323)
oven 50 °C (2 min), 4 °C/min to 210 °C
carrier gas helium, 25 cm/sec
sample Supelco 37-Component FAME Mix (47885-U), analytes at concentrations indicated in methylene chloride
injection 1 µL, 100:1 split
inj. temp. 250 °C
detector FID, 260 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages