GC Analysis of a 37-Component FAME Mix on SP™-2560 (75 m x 0.18 mm I.D., 0.14 μm), Fast GC Analysis
GC Analysis of a 37-Component FAME Mix on SP™-2560 (75 m x 0.18 mm I.D., 0.14 μm), Fast GC Analysis
Conditions
column SP-2560, 75 m x 0.18 mm I.D., 0.14 µm (23348-U)
oven 140 C (5 min), 4 C/min to 240 C (2 min)
carrier gas hydrogen, 40 cm/sec. @ 175 C
sample 37 component FAME mix at concentrations listed in methylene chloride (47885-U)
injection 1 µL, split 100:1
liner 4 mm I.D split, cup design
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information SP is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes