GC Analysis of a 37-Component FAME Mix on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of a 37-Component FAME Mix on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column Equity-1, 15 m x 0.10 mm I.D., 0.10 µm (28039-U)
oven 100 °C, 50 °C/min to 300 °C (1 min)
carrier gas hydrogen, 50 cm/sec constant
sample Supelco 37-Component FAME Mix (47885-U), analytes at concentrations indicated in methylene chloride
injection 0.2 µL, 200:1 split
liner 4 mm I.D., split, cup design
inj. temp. 250 °C
detector FID, 300 °C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • cis-11,14,17-Eicosatrienoic
  • cis-4,7,10,13,16,19-Docosahexaenoic
  • cis-8,11,14-Eicosatrienoic
  • Methyl all-cis-5,8,11,14,17-eicosapentaenoate
  • Methyl arachidate
  • Methyl arachidonate
  • Methyl behenate
  • Methyl butyrate
  • Methyl caprylate
  • Methyl cis,cis-11,14-eicosadienoate
  • Methyl cis-10-heptadecenoate
  • Methyl cis-10-pentadecenoate
  • Methyl cis-11-eicosenoate
  • Methyl cis-13,16-docosadienoate
  • Methyl cis-15-tetracosenoate
  • Methyl decanoate
  • Methyl elaidate
  • Methyl erucate
  • Methyl heneicosanoate
  • Methyl heptadecanoate
  • Methyl hexanoate
  • Methyl lactate
  • Methyl linoleate
  • Methyl linolelaidate
  • Methyl linolenate
  • Methyl myristate
  • Methyl myristoleate
  • Methyl oleate
  • Methyl palmitate
  • Methyl palmitoleate
  • Methyl pentadecanoate
  • Methyl stearate
  • Methyl tetracosanoate
  • Methyl tricosanoate
  • Methyl tridecanoate
  • Methyl undecanoate
  • Methyl γ-linolenate