GC Analysis of a 37-Component FAME Mix on SLB®-IL111
GC Analysis of a 37-Component FAME Mix on SLB®-IL111
Conditions
column SLB-IL111, 100 m x 0.25 mm I.D., 0.20 µm (29647-U)
oven 140 C (5 min), 8 C/min to 180 C, 5 C/min to 260 C
carrier gas hydrogen, 40 cm/sec
sample Supelco 37-Component FAME Mix (47885-U), analytes at concentrations indicated in methylene chloride
injection 1 µL, 100:1 split
liner 4 mm I.D., cup design
inj. temp. 250 C
detector FID, 260 C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes