Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of a 56-Component Industrial Solvents Mix on SLB®-IL60, Comparison to a Polyethylene Glycol (PEG) Column
GC Analysis of a 56-Component Industrial Solvents Mix on SLB®-IL60, Comparison to a Polyethylene Glycol (PEG) Column
Conditions
column SLB-IL60, 30 m x 0.25 mm I.D., 0.20 µm (29505-U)
column PEG 1, 30 m x 0.25 mm I.D., 0.25 µm
oven 40 C (4 min), 8 C/min to 200 C (5 min)
carrier gas helium, 30 cm/sec
sample 56-component industrial solvent mix, each analyte at 0.2% (v/v) in pentane
injection 1.0 muL100:1 split
liner 4 mm I.D., split/splitless type, single taper wool packed FocusLiner(TM) design
inj. temp. 250 C
detector FID, 250 C
Description
Categories Analytical Chromatography, GC Applications, Solvents
Featured Industry Chemicals and Industrial Polymers
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SLB is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 1,1,1,2-Tetrachloroethane
  • 1,1,1-Trichloroethane
  • 1,1,2,2-Tetrachloroethane
  • 1,1-Dichloroethane
  • 1,1-Dichloroethylene
  • 1,2-Dichloroethane
  • 1,4-Dichlorobenzene
  • 1,4-Dioxane
  • 2-Butanone
  • 2-Butoxyethanol (Butyl Cellosolve)
  • 2-Hexanone
  • 2-Methylphenol
  • 2-Propanol
  • 3-Methylphenol
  • 4-Methyl-2-pentanone
  • 4-Methylphenol
  • 5-Methyl-2-hexanone
  • Acetone
  • Benzene
  • Carbon tetrachloride
  • Chlorobenzene
  • Chloroform
  • Cyclohexanol
  • Diacetone alcohol
  • Dimethylformamide (DMF)
  • Ethanol
  • Ethyl acetate
  • Ethyl formate
  • Ethylbenzene
  • Hexane
  • Isoamyl acetate
  • Isoamyl alcohol
  • Isobutanol
  • Isobutyl acetate
  • Isopropyl acetate
  • Mesityl oxide
  • Methanol
  • Methyl acetate
  • Methyl formate
  • Methylene chloride
  • m-Xylene
  • n-Amyl acetate
  • n-Butanol
  • n-Butyl acetate
  • Nitropropane
  • n-Propanol
  • n-Propyl acetate
  • o-Xylene
  • p-Xylene
  • sec-Butanol
  • Styrene
  • Tetrachloroethene
  • Tetrahydrofuran (THF)
  • Toluene
  • trans-1,2-Dichloroethylene
  • Trichloroethylene