GC Analysis of a 6-Component Tetrachlorodibenzo-p-dioxin (TCDD) Mix on SP™-2331
GC Analysis of a 6-Component Tetrachlorodibenzo-p-dioxin (TCDD) Mix on SP™-2331
Conditions
column SP-2331, 60 m x 0.25 mm I.D., 0.20 µm (24104-U)
oven 170 °C (1 min), 8 °C/min to 265 °C
carrier gas helium, 37 cm/sec constant
sample 1.5 µg/mL TCDD standard in dodecane
injection 1 µLsplitless (1 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
MSD interface 265 °C
scan range SIM
Description
Categories Analytical Chromatography, GC Applications, Dioxins/PCBs/PBDEs
Featured Industry Agriculture
Environmental
Legal Information SP is a trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 1,2,3,4-Tetrachlorodibenzo-p-dioxin
  • 1,2,3,7-Tetrachlorodibenzo-p-dioxin
  • 1,2,3,8-Tetrachlorodibenzo-p-dioxin
  • 1,2,7,8-Tetrachlorodibenzo-p-dioxin
  • 1,4,7,8-Tetrachlorodibenzo-p-dioxin
  • 2,3,7,8-Tetrachlorodibenzo-p-dioxin solution