GC Analysis of a Grain Fatty Acid Methyl Ester (FAME) Mix on SP™-2560
GC Analysis of a Grain Fatty Acid Methyl Ester (FAME) Mix on SP™-2560
Conditions
column SP-2560, 100 m x 0.25 mm I.D., 0.20 mum film (24056)
oven 140 C (5 min) to 240 C at 4 C/min
carrier gas helium, 20 cm/sec
injection 1 µLsplit 100:1
inj. temp. 260 °C
detector FID, 260 C
Description
Categories Analytical Chromatography, GC Applications, Fatty Acid Methyl Esters (FAMEs)
Featured Industry Agriculture
Clinical
Food and Beverages
Legal Information SP is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Arachidic acid (C20:0)
  • Behenic acid (C22:0)
  • Capric acid (C10:0)
  • Caprylic acid (C8:0)
  • cis-11-Eicosenoate (C20:1)
  • Elaidic acid (C18:1n9t)
  • Erucic (C22:1n9)
  • Heptadecanoic acid (C17:0)
  • Lauric acid (C12:0)
  • Linoleic acid (C18:2n6c)
  • Linolenic acid (C18:3n3)
  • Myristic acid (C14:0)
  • Myristoleic acid (C14:1n9c)
  • Oleic acid (C18:1n9c)
  • Palmitic acid (C16:0)
  • Palmitoleic acid (C16:1n9c)
  • Pentadecanoic acid (C15:0)
  • Stearic acid (C18:0)
  • Tridecanoic acid (C13:0)