GC Analysis of a QC Test Mix Before/After Repeat 60%Water Injections on Watercol™ 1910
GC Analysis of a QC Test Mix Before/After Repeat 60%Water Injections on Watercol™ 1910
Conditions
column Watercol 1910, 30 m x 0.25 mm I.D., 0.20 µm (29711-U)
oven 163 °C
carrier gas helium, 26 cm/sec
sample 6 analytes in acetone, each at 1 mg/mL except water (at 0.5 mg/mL)
injection 0.5 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector TCD, 200 °C
Description
Analysis Note The "Watercol" series of ionic liquid capillary GC columns are characterized by their ability to produce a sharp peak shape for water, sharp enough so that water can be integrated and quantified. Sharp peak shapes are also produced for other small polar analytes. This application shows the ruggedness of Watercol 1910 to repeat injections of 60% water.
Featured Industry Chemicals and Industrial Polymers
Cosmetics, Personal Care, and Cleaning Products
Flavors and Fragrances
Food and Beverages
Petroleum
Pharmaceutical (small molecule)
Legal Information Watercol is a trademark of Sigma-Aldrich Co. LLC
Other Notes Conditions for each 60% water run = oven: 80 °C; inj. temp.: 250 °C; detector: TCD, 200 °C; carrier gas: helium, 28 cm/sec; injection: 1 μL, 100:1 split; liner: 4 mm I.D., split type, cup design; sample: water at 60% (v/v) in ethanol