GC Analysis of a QC Test Mix Containing Water on Watercol 1900, 96 °C
GC Analysis of a QC Test Mix Containing Water on Watercol 1900, 96 °C
Conditions
column Watercol 1900, 30 m x 0.25 mm I.D., 0.20 µm (29761-U)
oven 96 C
carrier gas helium, 26 cm/sec
sample 6 analytes in acetone, each at 1 mg/mL except water (at 0.5
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector TCD, 200 °C
Description
Analysis Note The "Watercol" series of ionic liquid capillary GC columns are characterized by their ability to produce a sharp peak shape for water, sharp enough so that water can be integrated and quantified. Sharp peak shapes are also produced for other small polar analytes. This application shows a QC test mix containing water on Watercol 1900 at 96 °C isothermal.
Featured Industry Chemicals and Industrial Polymers
Cosmetics, Personal Care, and Cleaning Products
Flavors and Fragrances
Food and Beverages
Petroleum
Pharmaceutical (small molecule)