GC Analysis of a QC Test Mix on SLB®-IL76i
GC Analysis of a QC Test Mix on SLB®-IL76i
Conditions
column SLB-IL76i, 30 m x 0.25 mm I.D., 0.20 µm (29869-U)
oven 130 °C
carrier gas helium, 1.2 mL/min (28 cm/sec)
sample polar column test mix (47302), 9 analytes, each at 500 µg/mL in methylene chlorid
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note The i-series of ionic liquid capillary GC columns are characterized by their improved inertness for polar analytes. This application shows that the SLB IL76i can generate great peak shapes for analytes with various functionality.
Featured Industry General Analytical
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC