GC Analysis of Active Amyl Alcohol, Isoamyl Alcohol, and C10-C14 n-Alkane Markers on SLB®-IL60i
GC Analysis of Active Amyl Alcohol, Isoamyl Alcohol, and C10-C14 n-Alkane Markers on SLB®-IL60i
Conditions
column SLB-IL60i, 30 m x 0.25 mm I.D., 0.20 µm (29832-U)
oven 90 °C
carrier gas hydrogen, 40 cm/sec
sample 7 analytes in methylene chloride
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Active amyl alcohol and isoamyl alcohol are by-products of the fermentation process. They are of concern to alcoholic beverage manufacturers due to their aroma characteristics. In this application, both analytes and several n-alkane markers are analyzed on SLB IL60i. This inert ionic liquid column produces great peak shapes for both alcohols, resulting in complete resolution.
Featured Industry Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC