GC Analysis of Active Amyl Alcohol, Isoamyl Alcohol, and C12-C14 n-Alkane Markers on SLB®-IL111i
GC Analysis of Active Amyl Alcohol, Isoamyl Alcohol, and C12-C14 n-Alkane Markers on SLB®-IL111i
Conditions
column SLB-IL111i, 30 m x 0.25 mm I.D., 0.20 µm (29883-U)
oven 90 °C
carrier gas hydrogen, 40 cm/sec
sample 5 analytes in methylene chloride
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup designdesign
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note Active amyl alcohol and isoamyl alcohol are by-products of the fermentation process. They are of concern to alcoholic beverage manufacturers due to their aroma characteristics. In this application, both analytes and several n-alkane markers are analyzed on SLB IL111i. This inert ionic liquic column produces great peak shapes for both alcohols, resulting in great resolution.
Featured Industry Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC