Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Agricultural Pesticides on SLB®-5ms
GC Analysis of Agricultural Pesticides on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D., 0.25 µm (28471-U)
oven 50 C (0.2 min), 100 C/min to 100 C, 5 C/min to 250 C, 15 C/min to 325 C (5 min)
carrier gas helium, 30 cm/sec constant
sample 10 ppm pesticides in ethyl acetate
injection 1 µL, splitless (0.75 min)
liner 2.6 mm I.D., straight
inj. temp. 250 C
MSD interface 300 C
scan range m/z = 40-450
Description
Categories Analytical Chromatography, Food & Beverage Analysis, GC Applications, Pesticide and Metabolite Residues, Herbicides, Applications (Chromatograms), Pesticides
Featured Industry Agriculture
Chemicals and Industrial Polymers
Environmental
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Acephate
  • Azoxystrobin
  • Benfluralin
  • Butamifos
  • Captan
  • Chlorofos (trichlorfan)
  • Chloroneb
  • Chlorothalonil
  • Chlorpyrifos
  • Diazinon
  • Dithiopyr
  • Ethofenprox
  • Etridiazol
  • Fenitrothion
  • Iprodione
  • Isofenphos
  • Isoprothiolane
  • Mecoprop methyl ester
  • Metalaxyl
  • Napropamide
  • Pendimethalin
  • Propiconazole isomers
  • Propyzamide
  • Pyridafenthion (pyridaphenthion)
  • Siduron isomers
  • Simazine
  • Terbucarb
  • Thiodicarb
  • Toclofos-methyl