GC Analysis of Agricultural Pesticides (Standard) on SLB®-5ms
GC Analysis of Agricultural Pesticides (Standard) on SLB®-5ms
Conditions
column SLB-5ms, 30 m x 0.25 mm I.D, 0.25 µm (28471-U)
oven 100 °C (1 min), 10 °C/min to 300 °C (5 min)
carrier gas helium, 1 mL/min constant
sample 29-component pesticide mix plus 1 internal standard, each at 100 ppb in toluene
injection 1.0 µLpulsed (20 psi until 0.20 min), splitless (1.0 min)
liner 4 mm I.D., single taper
inj. temp. 250 °C
MSD interface 300 °C
scan range selected ion monitoring (SIM), 7 monitoring groups used
Description
Analysis Note This application illustrates the analysis of 29 different agricultural pesticides of different classifications and functionalities. The analysis was done by GC-MS using selected ion monitoring (SIM) to allow detection of all compounds at 100 μg/L. The
standard solution was made in toluene to increase it stability and enhance response of some of the pesticides. The extra peaks in the first portion of the run are due to interferences from the solvent.
Categories Analytical Chromatography, GC Applications, Herbicides, Pesticides
Featured Industry Agriculture
Food and Beverages
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
suitability application for GC
Analyte
  • 4,4′-DDE (1,1-Dichloro-2,2-bis(p-chlorophenyl)ethylene)
  • Acephate
  • Captan
  • Carbaryl
  • Chlorothalonil
  • Chlorpyrifos
  • cis-Chlordane
  • cis-Permethrin
  • Coumaphos
  • Cyprodinil
  • Diazinon
  • Dichlofluanid
  • Dichlorvos
  • Dicofol
  • Dieldrin
  • Endosulfan sulfate
  • Ethoprophos
  • Folpet
  • Heptachlor epoxide
  • Hexachlorobenzene
  • Imazalil
  • Lindane
  • Methamidophos
  • Methyl chloropyrifos
  • Penconazole
  • Propoxur
  • Thiabendazole
  • Tolylfluanid
  • trans-Permethrin
  • γ-BHC