GC Analysis of Alcoholic Spirit on Equity®-1 Coupled to SUPELCOWAX 10 (Column Set 2)
GC Analysis of Alcoholic Spirit on Equity®-1 °Coupled to SUPELCOWAX 10 (Column Set 2)
Conditions
column Equity-1, 30 m x 0.32 mm I.D., 5.0 µm coupled to SUPELCOWAX 10, 10 m x 0.32 mm I.D., 0.25µm (28062-U)
oven 45 °C (1 min), 10 °C/min to 150 °C, 15 °C/min to 240 °C (2 min)
carrier gas helium, 36 cm/sec @ 45 °C
sample alcoholic spirit
injection 1 µL, 11:1 split
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Food & Beverage Analysis, Beverage Testing, Alcohols, Beer, Wine, and Spirits, Spirits
Featured Industry Food and Beverages
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1-Butanol
  • 1-Heptanol
  • 1-Hexanol
  • 1-Propanol
  • 2-Butanol
  • 2-Butanol
  • 2-Methyl-1-Propanol
  • 2-Propanol
  • 3-Methylbutanol
  • Acetaldehyde
  • Allyl alcohol
  • Diethyl succinate
  • Ethyl acetate
  • Ethyl lactate
  • Furfuryl alcohol
  • Isopentyl acetate
  • Methanol