GC Analysis of Alcohols and Hydrocarbons on Equity®-1
GC Analysis of Alcohols and Hydrocarbons on Equity®-1
Conditions
column Equity-1, 30 m x 0.53 mm I.D., 3.0 µm (28076-U)
oven 40 °C (5 min), 8 ºC/min to 225 °C (10 min)
carrier gas helium, 30 cm/sec @ 40 °C
sample 32 component mixed solvent sample, equal by weight
injection 0.1 µL, split 100:1
liner split, cup design
inj. temp. 250 °C
detector FID, 275 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons, Alcohols
Featured Industry Chemicals and Industrial Polymers
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes