GC Analysis of Allergens in Perfume on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Allergens in Perfume on SLB®-5ms (10 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column SLB-5ms, 10 m x 0.10 mm I.D., 0.10 µm (28465-U)
oven 40 °C, 50 °C/min to 320 °C
carrier gas hydrogen, 81.5 cm/sec constant
sample neat perfume
injection 0.2 µL in the split mode (500:1)
inj. temp. 320 °C
detector FID, 320 °C
Description
Categories Analytical Chromatography, GC Applications, Allergens
Featured Industry Cosmetics, Personal Care, and Cleaning Products
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • ®-(+)-Limonene
  • Cinnamyl alcohol
  • Citronellol
  • Coumarin
  • Eugenol
  • Geranial
  • Hexyl cinnamylaldehyde
  • Hydroxycitronellal
  • Linalool
  • Neral
  • α-Isomethyl ionone