GC Analysis of Amphetamines in Plasma on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm) after SPME using 100 μm PDMS Fiber, Fast GC Analysis
GC Analysis of Amphetamines in Plasma on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm) after SPME using 100 μm PDMS Fiber, Fast GC Analysis
Conditions
extraction headspace, 300 rpm stirring at 75 C for 15 min
SPME fiber 100 µm PDMS (57300-U)
column SLB-5ms, 20 m x 0.18 mm I.D. x 0.36 µm (28576-U)
oven 75 C (1 min), 10 C/min to 190C, 25 C/min to 300 C (4 min)
carrier gas helium, 0.7 ml/min. constant
liner 0.75 mm I.D. SPME
MSD interface 300 C
Description
Categories Analytical Chromatography, SPME Applications, GC Applications, Clinical, Forensic, Bioanalytical, Drugs & Pharmaceuticals, Drugs of Abuse, Amines, Pharmaceuticals
Featured Industry Clinical
Forensics and Toxicology
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for SPME, application for GC