Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of an 8-Component Solvent Mix in Water on Watercol 1460
GC Analysis of an 8-Component Solvent Mix in Water on Watercol 1460
Conditions
column Watercol 1460, 30 m x 0.25 mm I.D., 0.20 µm (29695-U)
oven 35 C, 4 C/min to 125 C (2 min)
carrier gas helium, 25 cm/sec
sample 8-component solvent mix in water
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup desig
inj. temp. 250 C
detector TCD, 300 C
Description
Analysis Note The "Watercol" series of ionic liquid capillary GC columns are characterized by their ability to produce a sharp peak shape for water, sharp enough so that water does not interfere chromatographically with other small polar analytes. This application shows the analysis of an 8-component solvent mix in water on Watercol 1460.
Featured Industry Chemicals and Industrial Polymers
Chemicals and Industrial Polymers
Environmental
Food and Beverages