GC Analysis of an 8-Component Solvent Mix in Water on Watercol™ 1900
GC Analysis of an 8-Component Solvent Mix in Water on Watercol™ 1900
Conditions
column Watercol 1900, 30 m x 0.25 mm I.D., 0.20 µm (29761-U)
oven 35 °C, 4 °C/min to 125 °C (2 min)
carrier gas helium, 25 cm/sec
sample 8-component solvent mix in water
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector TCD, 300 °C
Description
Analysis Note The "Watercol" series of ionic liquid capillary GC columns are characterized by their ability to produce a sharp peak shape for water, sharp enough so that water does not interfere chromatographically with other small polar analytes. This application shows the analysis of an 8-component solvent mix in water on Watercol 1900.
Featured Industry Chemicals and Industrial Polymers
Cosmetics, Personal Care, and Cleaning Products
Environmental
Food and Beverages
Legal Information Watercol is a trademark of Sigma-Aldrich Co. LLC