GC Analysis of an Indoor Air Standard on Equity®-1
GC Analysis of an Indoor Air Standard on Equity®-1
Conditions
column Equity®-1, 30 m x 0.25 mm I.D., 1.0 µm (28049-U)
oven 35 °C (3 min), 8 °C/min to 100 °C, 20 °C/min to 250 °C (10 min)
carrier gas helium, 30 cm/sec @ 35 °C
sample 100 ng on-column of the Japanese Indoor Air Standards Mix (47537-U)
injection 1 µL, 10:1 split
liner split, cup design
inj. temp. 250 °C
MSD interface 325 °C
scan range m/z 33-350
Description
Categories Analytical Chromatography, GC Applications, Air Monitoring Applications, Volatiles
Featured Industry Environmental
Industrial Hygiene
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for air monitoring, application for GC
Analyte
  • 1,1,1-Trichloroethane
  • 1,2,3-Trimethylbenzene
  • 1,2,4,5-Tetramethylbenzene
  • 1,2,4-Trimethylbenzene
  • 1,2-Dichloroethane
  • 1,2-Dichloropropane
  • 1,3,5-Trimethylbenzene
  • 1,4-Dichlorobenzene
  • 1-Butanol
  • 1-Propanol
  • 2,4-Dimethylpentane
  • 2-Butanone
  • 2-Ethyltoluene
  • 2-Propanol
  • 3-Ethylbenzene
  • 4-Ethylbenzene
  • 4-Methyl-2-pentanone
  • Acetone
  • Benzene
  • Bromodichloromethane
  • Carbon tetrachloride
  • Chlorodibromomethane
  • Chloroform
  • Decanal
  • Decane
  • Dodecane
  • Ethanol
  • Ethyl acetate
  • Ethylbenzene
  • Heptane
  • Hexadecane
  • Hexane
  • Isooctane
  • Limonene
  • Methylene chloride
  • m-Xylene
  • n-Butyl acetate
  • Nonanal
  • Nonane
  • Octane
  • o-Xylene
  • Pentadecane
  • p-Xylene
  • Styrene
  • Tetrachloroethylene
  • Tetradecane
  • Toluene
  • Trichloroethene
  • Tridecane
  • Undecane
  • α-Pinene
  • β-Pinene