GC Analysis of Antioxidants on SAC™-5
GC Analysis of Antioxidants on SAC™-5
Conditions
column SAC-5, 30 m x 0.25 mm I.D., 0.25 µm (24156)
oven 200 °C
carrier gas helium, 30 cm/sec
sample 200 µg/mL each component
injection 2 µL, split 100:1
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Antioxidants
Featured Industry Cosmetics, Personal Care, and Cleaning Products
Food and Beverages
Legal Information SAC is a trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • BHA
  • BHT
  • Ethoxyquin
  • Ionox 100
  • Propyl gallate
  • TBHQ
  • THBP