GC Analysis of Aromatics and Alcohols on SLB®-ILD3606, Isothermal Analysis
GC Analysis of Aromatics and Alcohols on SLB®-ILD3606, Isothermal Analysis
Conditions
column SLB-ILD3606, 60 m x 0.25 mm I.D., 0.20 µm (29691-U)
oven 50 °C (20 min)
carrier gas helium, 21 cm/sec
sample 8 analytes at various concentrations in isooctane
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup design
inj. temp. 250 °C
detector FID, 250 °C
Description
Analysis Note The measurement of aromatic and oxygenate compounds in petroleum samples is a common application performed worldwide, both in on-site labs at industrial facilities and also in third-party testing labs. The SLB-ILD3606 column is able to resolve benzene (and other aromatics) from oxygenates, such as alcohols.
Categories Analytical Chromatography, GC Applications, Aromatics, Alcohols, Ketones, Hydrocarbons
Featured Industry Petroleum
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC