GC Analysis of Aromatics and Alcohols on SLB®-IL111i, Temperature Programmed Analysis
GC Analysis of Aromatics and Alcohols on SLB®-IL111i, Temperature Programmed Analysis
Conditions
column SLB-IL111i, 60 m x 0.25 mm I.D., 0.20 µm (29884-U)
oven 50 °C (6 min), 15 °C/min to 265 °C (10 min)
carrier gas helium, 21 cm/sec
sample 8 analytes at various concentrations in isooctane
injection 1 µL, 100:1 split
liner 4 mm I.D., split type, cup design (2879925-U)
inj. temp. 250 °C
detector FID, 250
Description
Analysis Note The inert SLB IL111i ionic liquic column produces great peak shapes for alcohols via a temperature programmed analysis, providing resolution in a mix of aromatics. This is impressive for such a polar column.
Featured Industry Chemicals and Industrial Polymers
Petroleum
Legal Information SLB is a registered trademark of Sigma-Aldrich Co. LLC