GC Analysis of Aromatics in Reformulated Gasoline on Equity®-1
GC Analysis of Aromatics in Reforµlated Gasoline on Equity®-1
Conditions
column Equity-1, 60 m x 0.25 mm I.D., 0.25 µm (28047-U)
oven 50 C (3 min), 15 C/min to 265 C (5 min)
carrier gas helium, 30 cm/sec
sample reforµlated unleaded gasoline
injection 0.5 µL, 200:1 split
liner 4 mm I.D., splitless type, single taper wool packed FocusLiner(TM) design
inj. temp. 250 C
detector FID, 265 C
Description
Analysis Note This application illustrates the analysis of a sample of reforµlated gasoline on the Equity-1 column. Due to the nonpolar nature of the phase, there is a great deal of overlap in the elution ranges of the aliphatic and aromatic portions of this highly complex sample. Benzene is not baseline resolved even on a 60 m column. For this reason, this phase could not be used by itself for the analysis of benzene in this sample.
Categories Analytical Chromatography, GC Applications, Aromatics, Hydrocarbons
Featured Industry Petroleum
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,2,3-Trimethylbenzene
  • 1,2,4,5-Tetramethylbenzene
  • 1,2,4-Trimethylbenzene
  • 1-Methylnaphthalene
  • 2-Methylnaphthalene
  • Benzene
  • Decane
  • Ethanol
  • Ethylbenzene
  • Heptane
  • Hexane
  • m-Xylene
  • Naphthalene
  • Nonane
  • Octane
  • o-Xylene
  • Pentane
  • p-Xylene
  • Toluene
  • Undecane