Attention:
Certain features of Sigma-Aldrich.com will be down for maintenance the evening of Friday August 18th starting at 8:00 pm CDT until Saturday August 19th at 12:01 pm CDT.   Please note that you still have telephone and email access to our local offices. We apologize for any inconvenience.
GC Analysis of Aromatics in Reformulated Gasoline on SUPELCOWAX® 10
GC Analysis of Aromatics in Reforµlated Gasoline on SUPELCOWAX® 10
Conditions
column SUPELCOWAX 10, 60 m x 0.25 mm I.D., 0.25 µm (24081)
oven 50 C (3 min), 15 C/min to 265 C (5 min)
carrier gas helium, 30 cm/sec
sample reforµlated unleaded gasoline
injection 0.5 µL, 200:1 split
liner 4 mm I.D., splitless type, single taper wool packed FocusLiner(TM) design
inj. temp. 250 C
detector FID, 265 C
Description
Analysis Note This application illustrates the analysis of a sample of reforµlated gasoline on the SUPELCOWAX 10 column. Due to the polar nature of the phase, the aliphatics elute earlier than the aromatics. There is some overlap in the elution ranges, however it is not as severe as that exhibited by a nonpolar column.
Categories Analytical Chromatography, GC Applications, Aromatics, Hydrocarbons
Featured Industry Petroleum
Legal Information FocusLiner is a trademark of SGE Analytical Science Pty Ltd., SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • 1,2,3-Trimethylbenzene
  • 1,2,4,5-Tetramethylbenzene
  • 1,2,4-Trimethylbenzene
  • 1-Methylnaphthalene
  • 2-Methylnaphthalene
  • Benzene
  • C5-C9 Alkanes
  • Decane
  • Dodecane
  • Ethanol
  • Ethylbenzene
  • m-Xylene
  • Naphthalene
  • o-Xylene
  • p-Xylene
  • Tetradecane
  • Toluene
  • Tridecane
  • Undecane