GC Analysis of Aviation Gasoline on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Aviation Gasoline on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column Equity-1, 15 m x 0.10 mm I.D., 0.10 µm (28039-U)
oven 40 °C (1 min), 45 °C/min to 225 °C
carrier gas hydrogen, 45 cm/sec constant
sample Aviation gasoline, neat
injection 1 µL300:1 split
liner 2 mm I.D., straight
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Hydrocarbons
Featured Industry Petroleum
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes