GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Conditions
column Equity-1, 15 m x 0.10 mm I.D., 0.10 µm (28039-U)
oven 175 C, 30 C/min to 275 C (1 min)
carrier gas hydrogen, 45 cm/sec constant
sample Bacterial Acid Methyl Ester (BAME) Mix (47080-U), methyl ester derivatives, total concentration of 10 mg/mL in methyl caproate
injection 0.5 µL, 200:1 split
liner 4 mm I.D., split, cup design
inj. temp. 280 C
detector FID, 280 C
Description
Categories Analytical Chromatography, GC Applications, Bacterial Acid Methyl Esters (BAMEs)
Featured Industry Clinical
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • Methyl (±)-2-hydroxydecanoate
  • Methyl (±)-3-hydroxydodecanoate
  • Methyl 12-methyltetradecanoate
  • Methyl 13-methyltetradecanoate
  • Methyl 14-methylpentadecanoate
  • Methyl 15-methylhexadecanoate
  • Methyl 2-hydroxydodecanoate
  • Methyl 2-hydroxyhexadecanoate
  • Methyl 2-Hydroxytetradecanoate
  • Methyl 3-hydroxytetradecanoate
  • Methyl arachidate
  • Methyl cis-9-hexadecenoate
  • Methyl dodecanoate
  • Methyl elaidate
  • Methyl heptadecanoate
  • Methyl myristate
  • Methyl nonadecanoate
  • Methyl oleate
  • Methyl palmitate
  • Methyl pentadecanoate
  • Methyl stearate
  • Methyl tridecanoate
  • Methyl undecanoate
  • Methyl cis-15-tetracosenoate
  • Methyl cis-9,10-methylenehexadecanoate
  • Methyl cis-9,10-methyleneoctadecanoate