GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on SUPELCOWAX® 10
GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on SUPELCOWAX® 10
Conditions
column SUPELCOWAX 10, 30 m x 0.32 mm I.D., 0.25 µm (24082)
oven 50C (1 min), 5 °C/min to 225 °C (5 min)
carrier gas helium, 25 cm/sec (set at 200 °C)
sample bacterial methyl ester standards in methyl caproate, 10 mg/mL (47080-U)
injection 1 µL100:1 split
liner 4 mm I.D., cup style
inj. temp. 250 °C
detector FID, 250 °C
Description
Categories Analytical Chromatography, GC Applications, Bacterial Acid Methyl Esters (BAMEs)
Featured Industry Clinical
Legal Information SUPELCOWAX is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes