GC Analysis of Basic Drugs on Equity®-5
GC Analysis of Basic Drugs on Equity®-5
Conditions
column Equity-5, 30 m x 0.25 mm I.D., 0.25 µm (28089-U)
oven 45 °C (2 min), 25 °C/min to 110 °C, 15 °C/min to 200 °C, 6 C/min to 280 °C (3 min)
carrier gas helium, 40 psi for 0.2 min then 0.7 mL/min constant flow
sample ~15 ng on-column of a 16-component drug standard
injection 0.3 µL pulsed splitless @ 50 mL/min (0.5 min)
liner 2 mm I.D., splitless
inj. temp. 250 °C
MSD interface 325 °C
scan range m/z = 40-450
Description
Categories Analytical Chromatography, GC Applications, Drugs of Abuse, Pharmaceuticals
Featured Industry Forensics and Toxicology
Legal Information Equity is a registered trademark of Sigma-Aldrich Co. LLC
Other Notes Complementary products:
Syringes
Vials
Gas Purification and Gas Management
Inlet Liners, Glass Wool, and Specialized Hand Tools
Septa and Specialized Hand Tools
Column Ferrules, Nuts, and Specialized Hand Tools
suitability application for GC
Analyte
  • (±)-Methadone
  • (±)-Nicotine
  • Amitriptyline
  • Amphetamine
  • Caffeine
  • Cocaine
  • Codeine
  • Desipramine
  • Diazepam
  • Diphenhydramine
  • Fentanyl
  • Heroin
  • Lidocaine
  • Methamphetamine
  • Morphine
  • Phenobarbital